Assay Details
Target Gene Details
Entrez Gene ID: | 440925 |
Gene Name: | long intergenic non-protein coding RNA 1124 |
Gene Aliases: |
- |
Location: |
Chr.2:170712439-170714567 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01124 | NR_027433.1 | 1 | 1227 | |
| AK123485.1 | 1 | 1227 | BAC85625.1 |
Target Gene Details
Entrez Gene ID: | 101926913 |
Gene Name: | uncharacterized LOC101926913 |
Gene Aliases: |
- |
Location: |
Chr.2:170700368-170770766 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926913 | NR_110185.1 | |||
| BM931568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26289 | Chr.2:170713380 - 170717552 on Build GRCh38 | Loss |
|
| dgv7158n54 | Chr.2:170712123 - 170717019 on Build GRCh38 | Loss |
|
| nsv821786 | Chr.2:170711270 - 170718197 on Build GRCh38 | Gain |
|

Genomic Map