Assay Details
Target Gene Details
Entrez Gene ID: | 57552 |
Gene Name: | neutral cholesterol ester hydrolase 1 |
Gene Aliases: |
AADACL1, NCEH |
Location: |
Chr.3:172630645-172711218 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCEH1 | NM_001146276.1 | 5 | 1252 | NP_001139748.1 |
| NM_001146277.1 | 5 | 1218 | NP_001139749.1 | |
| NM_001146278.1 | 4 | 999 | NP_001139750.1 | |
| NM_020792.4 | 5 | 1228 | NP_065843.3 | |
| AB037784.1 | 5 | 1061 | ||
| AK294811.1 | 5 | 1071 | ||
| AK295641.1 | 4 | 842 | ||
| AK299422.1 | 5 | 1100 | ||
| AK304253.1 | 5 | 1129 | ||
| AK316431.1 | 5 | 1123 | ||
| BC028734.1 | 5 | 1233 | AAH28734.2 | |
| BC047588.1 | 5 | 1077 | AAH47588.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471060 | Chr.3:172622817 - 172682165 on Build GRCh38 | Gain |
|
| dgv8730n54 | Chr.3:172604186 - 172680078 on Build GRCh38 | Gain |
|
| dgv627n27 | Chr.3:172621962 - 172680078 on Build GRCh38 | Gain |
|
| esv3598669 | Chr.3:172612012 - 172681673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map