Assay Details
Target Gene Details
Entrez Gene ID: | 100131347 |
Gene Name: | RAD52 motif containing 1 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.17:39057019-39081451 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100131347 | NR_036551.1 | |||
| AK095115.1 | ||||
| AK124220.1 | BAC85811.1 |
Target Gene Details
Entrez Gene ID: | 57125 |
Gene Name: | plexin domain containing 1 |
Gene Aliases: |
TEM3, TEM7 |
Location: |
Chr.17:39063303-39151649 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLXDC1 | NM_020405.4 | 14 | 5527 | NP_065138.2 |
| AF070526.1 | 1 | 1227 | ||
| AF378753.1 | 14 | 5397 | AAL11990.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833440 | Chr.17:39033205 - 39210437 on Build GRCh38 | Loss |
|
| esv3640537 | Chr.17:39059290 - 39066885 on Build GRCh38 | Loss |
|
| nsv2046 | Chr.17:39023022 - 39084760 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78151658] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map