Assay Details
Target Gene Details
Entrez Gene ID: | 55294 |
Gene Name: | F-box and WD repeat domain containing 7 |
Gene Aliases: |
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4 |
Location: |
Chr.4:152321258-152536095 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXW7 | NM_001013415.1 | 6 | 866 | NP_001013433.1 |
| NM_018315.4 | 6 | 1031 | NP_060785.2 | |
| NM_033632.3 | 7 | 1205 | NP_361014.1 | |
| XM_011532083.1 | 9 | 2218 | XP_011530385.1 | |
| XM_011532084.1 | 8 | 2168 | XP_011530386.1 | |
| XM_011532085.1 | 7 | 2075 | XP_011530387.1 | |
| XM_011532086.1 | 10 | 2396 | XP_011530388.1 | |
| XM_011532087.1 | 9 | 2346 | XP_011530389.1 | |
| XM_011532088.1 | 7 | 1604 | XP_011530390.1 | |
| XM_017008362.1 | 9 | 2615 | XP_016863851.1 | |
| AB209725.1 | 6 | 795 | BAD92962.1 | |
| AF383178.1 | 6 | 605 | AAK60269.1 | |
| AF411971.1 | 6 | 1118 | AAL06290.1 | |
| AF411972.1 | 6 | 899 | AAL06291.1 | |
| AK001933.1 | 6 | 656 | BAA91986.1 | |
| AK223449.1 | 6 | 833 | BAD97169.1 | |
| AK294568.1 | 5 | 592 | ||
| AK299722.1 | 5 | 614 | ||
| AK315142.1 | 6 | 813 | ||
| AY008274.1 | 6 | 868 | AAG16640.1 | |
| AY033553.1 | 6 | 899 | AAK57547.1 | |
| AY049984.1 | 6 | 1112 | AAL07271.1 | |
| BC037320.2 | 6 | 786 | AAH37320.1 | |
| BC117244.1 | 7 | 1173 | ||
| BC117246.1 | 7 | 1173 | ||
| BC143944.1 | 8 | 1223 | ||
| CR749305.1 | 7 | 1336 | CAH18160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4553 | Chr.4:152323453 - 152349980 on Build GRCh38 | Insertion |
|
| nsv516218 | Chr.4:152308716 - 152388386 on Build GRCh38 | Loss |
|
| dgv5440n100 | Chr.4:152321467 - 152392321 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map