Assay Details
Target Gene Details
Entrez Gene ID: | 26007 |
Gene Name: | triokinase and FMN cyclase |
Gene Aliases: |
DAK, NET45 |
Location: |
Chr.11:61333182-61353426 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TKFC | NM_015533.3 | 18 | 2059 | NP_056348.2 |
| XM_011544912.2 | XP_011543214.1 | |||
| XM_017017517.1 | 18 | 3277 | XP_016873006.1 | |
| XM_017017518.1 | XP_016873007.1 | |||
| XM_017017519.1 | XP_016873008.1 | |||
| XM_017017520.1 | 18 | 3333 | XP_016873009.1 | |
| XM_017017521.1 | 19 | 2121 | XP_016873010.1 | |
| XM_017017522.1 | XP_016873011.1 | |||
| XM_017017523.1 | 18 | 2090 | XP_016873012.1 | |
| XM_017017524.1 | 19 | 2200 | XP_016873013.1 | |
| AK023915.1 | BAB14722.1 | |||
| AL110260.1 | 5 | 574 | CAB53701.1 | |
| BC001341.1 | 17 | 1853 | AAH01341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428259 | Chr.11:61049513 - 61356164 on Build GRCh38 | Gain+Loss |
|
| nsv832177 | Chr.11:61185118 - 61356164 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map