Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map