Assay Details
Target Gene Details
Entrez Gene ID: | 22858 |
Gene Name: | intestinal cell (MAK-like) kinase |
Gene Aliases: |
ECO, LCK2, MRK |
Location: |
Chr.6:53001299-53061864 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICK | NM_014920.3 | NP_055735.1 | ||
| NM_016513.4 | NP_057597.2 | |||
| XM_011514419.1 | XP_011512721.1 | |||
| XM_011514420.2 | XP_011512722.1 | |||
| XM_011514421.1 | XP_011512723.1 | |||
| XM_017010485.1 | XP_016865974.1 | |||
| XM_017010486.1 | XP_016865975.1 | |||
| XM_017010487.1 | XP_016865976.1 | |||
| XM_017010488.1 | XP_016865977.1 | |||
| XM_017010489.1 | XP_016865978.1 | |||
| XM_017010490.1 | XP_016865979.1 | |||
| XM_017010491.1 | XP_016865980.1 | |||
| XM_017010492.1 | XP_016865981.1 | |||
| AB023153.1 | ||||
| AF152469.1 | AAG43364.1 | |||
| AF225919.1 | AAF37278.1 | |||
| AK074892.1 | ||||
| BC136420.1 | ||||
| BC136421.1 | ||||
| BC152464.1 |
More Information
Set Membership: |
|

Genomic Map