Assay Details
Target Gene Details
Entrez Gene ID: | 114825 |
Gene Name: | PWWP domain containing 2A |
Gene Aliases: |
MST101 |
Location: |
Chr.5:160043538-160120709 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PWWP2A | XM_011534424.2 | XP_011532726.1 | ||
| XM_017009012.1 | XP_016864501.1 |
Target Gene Details
Entrez Gene ID: | 7265 |
Gene Name: | tetratricopeptide repeat domain 1 |
Gene Aliases: |
TPR1 |
Location: |
Chr.5:160009100-160065545 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC1 | NM_001282500.1 | NP_001269429.1 | ||
| NM_003314.2 | NP_003305.1 | |||
| AK128840.1 | ||||
| AK222641.1 | BAD96361.1 | |||
| BC000942.3 | AAH00942.1 | |||
| BQ017702.1 | ||||
| U46570.1 | AAB36871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830531 | Chr.5:159896337 - 160076045 on Build GRCh38 | Loss |
|
| dgv1151e214 | Chr.5:160058935 - 160082069 on Build GRCh38 | Gain |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map