Assay Details
Target Gene Details
Entrez Gene ID: | 84628 |
Gene Name: | netrin G2 |
Gene Aliases: |
LHLL9381, Lmnt2, NTNG1, bA479K20.1 |
Location: |
Chr.9:132161234-132244534 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTNG2 | NM_032536.2 | 2 | 795 | NP_115925.2 |
| XM_006717304.3 | 4 | 816 | XP_006717367.1 | |
| XM_011519094.2 | 4 | 816 | XP_011517396.1 | |
| XM_011519096.2 | 4 | 1213 | XP_011517398.1 | |
| XM_011519097.2 | 3 | 734 | XP_011517399.1 | |
| XM_011519098.2 | 4 | 766 | XP_011517400.1 | |
| XM_011519099.2 | 3 | 737 | XP_011517401.1 | |
| XM_011519100.2 | 2 | 666 | XP_011517402.1 | |
| XM_011519102.2 | 3 | 681 | XP_011517404.1 | |
| XM_011519103.2 | 2 | 573 | XP_011517405.1 | |
| XM_011519104.2 | 2 | 797 | XP_011517406.1 | |
| XM_011519105.2 | 2 | 663 | XP_011517407.1 | |
| XM_011519106.2 | 4 | 816 | XP_011517408.1 | |
| XM_011519107.2 | 4 | 816 | XP_011517409.1 | |
| XM_011519108.2 | 4 | 816 | XP_011517410.1 | |
| XM_011519109.2 | 4 | 816 | XP_011517411.1 | |
| XM_011519110.2 | 4 | 816 | XP_011517412.1 | |
| XM_017015212.1 | 5 | 920 | XP_016870701.1 | |
| XM_017015213.1 | 3 | 1123 | XP_016870702.1 | |
| XM_017015214.1 | 4 | 816 | XP_016870703.1 | |
| XM_017015216.1 | 4 | 816 | XP_016870705.1 | |
| AB058760.2 | 2 | 795 | ||
| AY358165.1 | 3 | 365 | AAQ88532.1 | |
| BC013770.1 | 1 | 104 | AAH13770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519872 | Chr.9:132164169 - 132175336 on Build GRCh38 | Loss |
|
| nsv509328 | Chr.9:132084017 - 132270727 on Build GRCh38 | Insertion |
|
| nsv615571 | Chr.9:132118176 - 132180427 on Build GRCh38 | Loss |
|
| nsv1035219 | Chr.9:132147299 - 132243197 on Build GRCh38 | Gain |
|
| nsv6739 | Chr.9:132135179 - 132166954 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map