Assay Details
Target Gene Details
Entrez Gene ID: | 130951 |
Gene Name: | meiosis 1 associated protein |
Gene Aliases: |
C2orf65, D6Mm5e, SPATA37 |
Location: |
Chr.2:74557640-74648338 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| M1AP | NM_001281296.1 | 10 | 1886 | NP_001268225.1 |
| NM_001321739.1 | 11 | 1898 | NP_001308668.1 | |
| NM_138804.4 | 11 | 1889 | NP_620159.2 | |
| XM_005264152.3 | 6 | 1046 | XP_005264209.1 | |
| XM_006711946.3 | 11 | 2234 | XP_006712009.1 | |
| XM_011532548.2 | 10 | 2223 | XP_011530850.1 | |
| XM_011532549.2 | 11 | 2176 | XP_011530851.1 | |
| XM_011532550.2 | 11 | 1845 | XP_011530852.1 | |
| XM_011532551.2 | 11 | 1844 | XP_011530853.1 | |
| XM_011532552.2 | 10 | 2061 | XP_011530854.1 | |
| XM_011532553.2 | 10 | 2029 | XP_011530855.1 | |
| XM_011532554.2 | 7 | 1058 | XP_011530856.1 | |
| AK126374.1 | 6 | 1047 | ||
| AK130161.1 | 6 | 862 | BAC85295.1 | |
| AK300188.1 | 10 | 1878 | ||
| BC025997.1 | 11 | 1892 | AAH25997.1 |
More Information
Set Membership: |
|

Genomic Map