Assay Details
Target Gene Details
Entrez Gene ID: | 1267 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNP1 |
Location: |
Chr.17:41966741-41977736 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNP | NM_033133.4 | 4 | 3920 | NP_149124.3 |
| AK098048.1 | 1 | 738 | ||
| AK124861.1 | 1 | 1270 |
Target Gene Details
Entrez Gene ID: | 7266 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C7 |
Gene Aliases: |
DJ11, DJC7, TPR2, TTC2 |
Location: |
Chr.17:41976421-42021371 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC7 | NM_001144766.2 | NP_001138238.1 | ||
| NM_003315.3 | NP_003306.3 | |||
| NR_029431.1 | ||||
| XM_011525167.2 | XP_011523469.1 | |||
| XM_011525168.2 | XP_011523470.1 | |||
| XM_011525169.2 | XP_011523471.1 | |||
| XM_017024994.1 | XP_016880483.1 | |||
| AB209834.1 | BAD93071.1 | |||
| AK291047.1 | ||||
| AK298860.1 | ||||
| BC003601.1 | AAH03601.1 | |||
| BC011837.2 | AAH11837.2 | |||
| BC033772.1 | AAH33772.1 | |||
| BQ445927.1 | ||||
| BX647209.1 | ||||
| U46571.1 | AAB36872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509658 | Chr.17:41927491 - 42002888 on Build GRCh38 | Insertion |
|
| nsv953897 | Chr.17:41966883 - 41977382 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map