Assay Details
Target Gene Details
Entrez Gene ID: | 11251 |
Gene Name: | prostaglandin D2 receptor 2 |
Gene Aliases: |
CD294, CRTH2, DL1R, DP2, GPR44 |
Location: |
Chr.11:60850925-60855971 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGDR2 | NM_004778.2 | NP_004769.2 | ||
| AB008535.1 | BAA74518.1 | |||
| AF144308.1 | AAD34539.1 | |||
| AK291371.1 | ||||
| AK313849.1 | ||||
| AY507142.1 | AAR92484.1 | |||
| BC096841.1 | AAH96841.1 |
More Information
Set Membership: |
|

Genomic Map