Assay Details
Target Gene Details
Entrez Gene ID: | 158584 |
Gene Name: | fatty acid amide hydrolase 2 |
Gene Aliases: |
AMDD |
Location: |
Chr.X:57121861-57489196 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAH2 | NM_174912.3 | NP_777572.2 | ||
| XM_005261982.1 | XP_005262039.1 | |||
| XM_005261985.2 | XP_005262042.1 | |||
| XM_005261986.3 | XP_005262043.1 | |||
| XM_005261987.3 | XP_005262044.1 | |||
| XM_011530767.2 | XP_011529069.1 | |||
| XM_017029290.1 | XP_016884779.1 | |||
| XM_017029291.1 | XP_016884780.1 | |||
| XM_017029292.1 | XP_016884781.1 | |||
| XM_017029293.1 | XP_016884782.1 | |||
| XM_017029294.1 | XP_016884783.1 | |||
| AK055766.1 | BAB71007.1 | |||
| BC048279.1 | AAH48279.1 | |||
| BC073922.1 | AAH73922.1 | |||
| DA392815.1 | ||||
| EU285560.1 |
More Information
Set Membership: |
|

Genomic Map