Assay Details
Target Gene Details
Entrez Gene ID: | 121642 |
Gene Name: | alkB homolog 2, alpha-ketoglutarate dependent dioxygenase |
Gene Aliases: |
ABH2 |
Location: |
Chr.12:109088188-109093631 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH2 | NM_001001655.2 | 2 | 437 | NP_001001655.1 |
| NM_001145374.1 | 2 | 587 | NP_001138846.1 | |
| NM_001145375.1 | 2 | 334 | NP_001138847.1 | |
| NM_001205179.1 | 2 | 484 | NP_001192108.1 | |
| NM_001205180.1 | 2 | 437 | NP_001192109.1 | |
| XM_005253835.4 | 2 | 603 | XP_005253892.1 | |
| XM_005253836.1 | 2 | 730 | XP_005253893.1 | |
| AB277859.1 | 1 | 200 | ||
| AK309062.1 | 2 | 573 | ||
| AY754389.1 | 2 | 422 | AAV28301.1 | |
| BC070489.1 | 2 | 389 | AAH70489.1 | |
| BM785502.1 | 2 | 437 | ||
| BM790974.1 | 2 | 316 | ||
| BM820131.1 | ||||
| BU623121.1 | 2 | 430 | ||
| CN421655.1 | 2 | 462 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1553n100 | Chr.12:109064624 - 109159352 on Build GRCh38 | Gain |
|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map