Assay Details
Target Gene Details
Entrez Gene ID: | 55216 |
Gene Name: | chromosome 11 open reading frame 57 |
Gene Aliases: |
- |
Location: |
Chr.11:112074244-112085150 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf57 | NM_001082969.1 | 6 | 829 | NP_001076438.1 |
| NM_001082970.1 | 6 | 1384 | NP_001076439.1 | |
| NM_001301017.1 | 6 | 826 | NP_001287946.1 | |
| NM_001301019.1 | 6 | 870 | NP_001287948.1 | |
| NM_001301021.1 | 6 | 867 | NP_001287950.1 | |
| NM_018195.3 | 6 | 1387 | NP_060665.3 | |
| NR_103469.1 | 6 | 1428 | ||
| AB096251.1 | 6 | 733 | ||
| AK001588.1 | 3 | 445 | BAA91774.1 | |
| AK125340.1 | 6 | 1340 | BAC86137.1 | |
| BC005403.1 | 4 | 626 | ||
| BC030546.1 | 4 | 631 | ||
| BC048133.1 | 6 | 1315 | ||
| BC072455.1 | 6 | 751 | ||
| BX538107.1 | 6 | 1386 | CAD98020.1 | |
| CD244552.1 | ||||
| EL736728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map