Assay Details
Target Gene Details
Entrez Gene ID: | 6376 |
Gene Name: | C-X3-C motif chemokine ligand 1 |
Gene Aliases: |
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin |
Location: |
Chr.16:57372461-57385048 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CX3CL1 | NM_001304392.1 | 2 | 3061 | NP_001291321.1 |
| NM_002996.4 | 3 | 3182 | NP_002987.1 | |
| AB209037.1 | 2 | 5664 | BAD92274.1 | |
| AK223351.1 | 5 | 1331 | BAD97071.1 | |
| BC001163.1 | 3 | 3141 | AAH01163.1 | |
| BC016164.1 | 3 | 3141 | AAH16164.1 | |
| GU047346.1 | 1 | 1883 | ||
| U84487.1 | 3 | 3153 | AAB49679.1 |
More Information
Set Membership: |
|

Genomic Map