Assay Details
Target Gene Details
Entrez Gene ID: | 145741 |
Gene Name: | C2 calcium dependent domain containing 4A |
Gene Aliases: |
FAM148A, NLF1 |
Location: |
Chr.15:62066977-62070917 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2CD4A | NM_207322.2 | 1 | 71 | NP_997205.2 |
| AF504646.1 | 1 | 45 | AAM28197.1 | |
| BM742013.1 | 1 | 71 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457178 | Chr.15:62047927 - 62102125 on Build GRCh38 | Loss |
|
| nsv1560 | Chr.15:62062065 - 62107105 on Build GRCh38 | Deletion |
|
| nsv457176 | Chr.15:61886496 - 62227459 on Build GRCh38 | Gain |
|
| nsv471247 | Chr.15:61910282 - 62227459 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map