Assay Details
Target Gene Details
Entrez Gene ID: | 102466736 |
Gene Name: | microRNA 6789 |
Gene Aliases: |
hsa-mir-6789 |
Location: |
Chr.19:2235829-2235926 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6789 | NR_106847.1 |
Target Gene Details
Entrez Gene ID: | 55111 |
Gene Name: | pleckstrin homology domain containing J1 |
Gene Aliases: |
GNRPX |
Location: |
Chr.19:2229952-2236724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHJ1 | NM_001300836.1 | NP_001287765.1 | ||
| NM_018049.2 | NP_060519.1 | |||
| XM_017026924.1 | XP_016882413.1 | |||
| XM_017026925.1 | XP_016882414.1 | |||
| XM_017026926.1 | XP_016882415.1 | |||
| XM_017026927.1 | XP_016882416.1 | |||
| XM_017026928.1 | XP_016882417.1 | |||
| AK001159.1 | BAA91525.1 | |||
| AK092742.1 | 2 | 311 | ||
| AK097723.1 | ||||
| AK098725.1 | ||||
| BC003084.2 | AAH03084.1 | |||
| BC008966.2 | AAH08966.1 | |||
| CN365243.1 | ||||
| CR457253.1 | CAG33534.1 | |||
| HY085955.1 | ||||
| HY101097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2389 | Chr.19:2206862 - 2240689 on Build GRCh38 | Insertion |
|
| nsv578382 | Chr.19:2194897 - 2260362 on Build GRCh38 | Loss |
|
| nsv953943 | Chr.19:2169602 - 2236001 on Build GRCh38 | Deletion |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv833706 | Chr.19:2131211 - 2249764 on Build GRCh38 | Loss |
|
| nsv578383 | Chr.19:2214058 - 2285197 on Build GRCh38 | Loss |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map