Assay Details
Target Gene Details
Entrez Gene ID: | 391059 |
Gene Name: | ferric chelate reductase 1 |
Gene Aliases: |
SDFR2, SDR2 |
Location: |
Chr.1:99708699-99766630 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRRS1 | NM_001013660.2 | 17 | 2382 | NP_001013682.2 |
| XM_005270861.3 | 17 | 1937 | XP_005270918.1 | |
| XM_011541451.2 | 17 | 2410 | XP_011539753.1 | |
| XM_017001271.1 | 15 | 1821 | XP_016856760.1 | |
| AK131302.1 | 17 | 2382 | BAD18470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002853 | Chr.1:99682647 - 99924231 on Build GRCh38 | Gain |
|
| nsv998891 | Chr.1:99479588 - 99843250 on Build GRCh38 | Loss |
|
| nsv526047 | Chr.1:99684466 - 99980490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map