Assay Details
Target Gene Details
Entrez Gene ID: | 8515 |
Gene Name: | integrin subunit alpha 10 |
Gene Aliases: |
PRO827 |
Location: |
Chr.1:145891208-145910189 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA10 | NM_001303040.1 | 27 | 4022 | NP_001289969.1 |
| NM_001303041.1 | 26 | 3910 | NP_001289970.1 | |
| NM_003637.4 | 30 | 4339 | NP_003628.2 | |
| XM_005277436.3 | 29 | 4090 | XP_005277493.1 | |
| XM_011510083.2 | 26 | 3770 | XP_011508385.1 | |
| XM_011510084.1 | 19 | 2767 | XP_011508386.1 | |
| XM_017002622.1 | 28 | 4296 | XP_016858111.1 | |
| XM_017002623.1 | 28 | 4293 | XP_016858112.1 | |
| XM_017002624.1 | 27 | 4182 | XP_016858113.1 | |
| XM_017002625.1 | 27 | 4182 | XP_016858114.1 | |
| XM_017002626.1 | 26 | 4053 | XP_016858115.1 | |
| XM_017002628.1 | 24 | 3746 | XP_016858117.1 | |
| AF112345.1 | 30 | 4240 | AAF21944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
| esv3587420 | Chr.1:145824369 - 145905171 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map