Assay Details
Target Gene Details
Entrez Gene ID: | 203260 |
Gene Name: | coiled-coil domain containing 107 |
Gene Aliases: |
PSEC0222 |
Location: |
Chr.9:35658282-35661503 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC107 | NM_001195200.1 | NP_001182129.1 | ||
| NM_001195201.1 | NP_001182130.1 | |||
| NM_001195217.1 | NP_001182146.1 | |||
| NM_174923.2 | NP_777583.2 | |||
| XM_005251403.4 | XP_005251460.1 | |||
| AK075523.1 | BAC11669.1 | |||
| AK307223.1 | 2 | 272 | ||
| BC018758.1 | AAH18758.1 | |||
| BC051285.1 | AAH51285.1 | |||
| BE903559.1 | ||||
| DC410699.1 | 2 | 347 | ||
| DQ884403.1 |

Genomic Map