Assay Details
Target Gene Details
Entrez Gene ID: | 604 |
Gene Name: | B-cell CLL/lymphoma 6 |
Gene Aliases: |
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51 |
Location: |
Chr.3:187721377-187745725 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL6 | NM_001130845.1 | 10 | 2534 | NP_001124317.1 |
| NM_001134738.1 | 8 | 1958 | NP_001128210.1 | |
| NM_001706.4 | 10 | 2442 | NP_001697.2 | |
| XM_005247694.3 | 10 | 5786 | XP_005247751.1 | |
| XM_011513062.2 | 9 | 2274 | XP_011511364.1 | |
| AK304325.1 | 10 | 2101 | ||
| AK308162.1 | 10 | 2183 | ||
| BC142705.1 | 10 | 2157 | ||
| BC150184.1 | 10 | 2158 | ||
| BX649185.1 | 10 | 3064 | CAE46203.1 | |
| EU139066.1 | 7 | 1933 | ||
| EU883531.1 | ||||
| S67779.1 | 10 | 2496 | ||
| U00115.1 | 10 | 2404 | AAC50054.1 | |
| Z21943.1 | 10 | 2161 | CAA79937.1 |
Target Gene Details
Entrez Gene ID: | 100131635 |
Gene Name: | hCG1645011-like |
Gene Aliases: |
- |
Location: |
Chr.3:187702366-187732415 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100131635 | NR_034062.1 | |||
| AW235368.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113181701] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map