Assay Details
Target Gene Details
Entrez Gene ID: | 11216 |
Gene Name: | A-kinase anchoring protein 10 |
Gene Aliases: |
AKAP-10, D-AKAP-2, D-AKAP2, PRKA10 |
Location: |
Chr.17:19904452-19977856 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP10 | NM_007202.3 | 15 | 2653 | NP_009133.2 |
| XM_006721432.3 | 14 | 2474 | XP_006721495.1 | |
| BC017055.1 | 15 | 2609 | AAH17055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76927580] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map