Assay Details
Target Gene Details
Entrez Gene ID: | 212 |
Gene Name: | 5'-aminolevulinate synthase 2 |
Gene Aliases: |
ALAS-E, ALASE, ANH1, ASB, SIDBA1, XLDPP, XLEPP, XLSA |
Location: |
Chr.X:55009055-55031064 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALAS2 | NM_000032.4 | 7 | 1105 | NP_000023.2 |
| NM_001037967.3 | 6 | 994 | NP_001033056.1 | |
| NM_001037968.3 | 7 | 1101 | NP_001033057.1 | |
| XM_005261995.3 | 8 | 1125 | XP_005262052.1 | |
| XM_011530771.1 | 5 | 684 | XP_011529073.1 | |
| XM_017029354.1 | 8 | 1126 | XP_016884843.1 | |
| AK290565.1 | 6 | 978 | ||
| AK291589.1 | 6 | 907 | ||
| AK313118.1 | 7 | 1105 | ||
| BC030230.2 | 6 | 980 | ||
| X56352.1 | 6 | 964 | CAA39795.1 | |
| X60364.1 | 7 | 1019 | CAA42916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map