Assay Details
Target Gene Details
Entrez Gene ID: | 2735 |
Gene Name: | GLI family zinc finger 1 |
Gene Aliases: |
GLI |
Location: |
Chr.12:57460135-57472268 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLI1 | NM_001160045.1 | NP_001153517.1 | ||
| NM_001167609.1 | NP_001161081.1 | |||
| NM_005269.2 | NP_005260.1 | |||
| XM_011538189.2 | XP_011536491.1 | |||
| XM_011538190.1 | XP_011536492.1 | |||
| AB239135.1 | ||||
| AB239136.1 | ||||
| AB239328.1 | ||||
| AK297899.1 | ||||
| BC013000.2 | AAH13000.1 | |||
| GQ890670.2 | ||||
| X07384.1 | CAA30297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759901 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv952190 | Chr.12:57459418 - 57466117 on Build GRCh38 | Deletion |
|
| dgv2651n54 | Chr.12:57450266 - 57552694 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv8974 | Chr.12:57458996 - 57472098 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map