Assay Details
Target Gene Details
Entrez Gene ID: | 5890 |
Gene Name: | RAD51 paralog B |
Gene Aliases: |
R51H2, RAD51L1, REC2 |
Location: |
Chr.14:67819779-68683118 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51B | NM_001321809.1 | NP_001308738.1 | ||
| NM_001321810.1 | NP_001308739.1 | |||
| NM_001321814.1 | NP_001308743.1 | |||
| NM_001321817.1 | NP_001308746.1 | |||
| NM_001321818.1 | NP_001308747.1 | |||
| NM_001321819.1 | NP_001308748.1 | |||
| NM_001321821.1 | NP_001308750.1 | |||
| NM_002877.5 | NP_002868.1 | |||
| NM_133509.3 | NP_598193.2 | |||
| NM_133510.3 | NP_598194.1 | |||
| XM_011537050.2 | XP_011535352.1 | |||
| XM_011537051.2 | XP_011535353.1 | |||
| XM_017021545.1 | XP_016877034.1 | |||
| AK309787.1 | ||||
| AK310686.1 | ||||
| AL597484.1 | ||||
| BC030219.1 | AAH30219.1 | |||
| BX161515.1 | CAD61950.1 | |||
| BX248061.1 | CAD62357.1 | |||
| BX248766.1 | CAD66573.1 | |||
| U84138.1 | AAC39723.1 | |||
| U92074.1 | AAB63358.1 | |||
| Y15571.1 | CAA75680.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054697 | Chr.14:67735287 - 67879265 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map