Assay Details
Target Gene Details
Entrez Gene ID: | 54972 |
Gene Name: | transmembrane protein 132A |
Gene Aliases: |
GBP, HSPA5BP1 |
Location: |
Chr.11:60924441-60937159 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM132A | NM_017870.3 | NP_060340.2 | ||
| NM_178031.2 | NP_821174.1 | |||
| XM_017017951.1 | XP_016873440.1 | |||
| XM_017017952.1 | XP_016873441.1 | |||
| AB046803.1 | 5 | 1356 | BAB13409.1 | |
| AK000546.1 | BAA91245.1 | |||
| AK023577.1 | BAB14613.1 | |||
| AL359557.2 | CAH10668.2 | |||
| AL834133.1 | ||||
| BC028106.1 | AAH28106.1 | |||
| BC032059.1 | AAH32059.1 | |||
| BC114569.1 | AAI14570.1 |
More Information
Set Membership: |
|

Genomic Map