Assay Details
Target Gene Details
Entrez Gene ID: | 654463 |
Gene Name: | fer-1 like family member 6 |
Gene Aliases: |
C8ORFK23 |
Location: |
Chr.8:123851987-124120061 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FER1L6 | NM_001039112.2 | NP_001034201.2 | ||
| XM_006716618.3 | XP_006716681.1 | |||
| XM_011517231.2 | XP_011515533.1 | |||
| XM_011517232.2 | XP_011515534.1 | |||
| XM_011517233.2 | XP_011515535.1 | |||
| XM_011517236.2 | XP_011515538.1 | |||
| AB196633.1 | BAE53435.1 |
Target Gene Details
Entrez Gene ID: | 157376 |
Gene Name: | FER1L6 antisense RNA 2 |
Gene Aliases: |
C8orf78 |
Location: |
Chr.8:124046071-124171522 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FER1L6-AS2 | NR_103547.1 | |||
| AK057332.1 | BAB71431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612144 | Chr.8:124045487 - 124194262 on Build GRCh38 | Loss |
|
| nsv1023183 | Chr.8:124048291 - 124194204 on Build GRCh38 | Loss |
|
| esv23245 | Chr.8:124089440 - 124098445 on Build GRCh38 | Loss |
|
| nsv831452 | Chr.8:124087467 - 124247685 on Build GRCh38 | Gain |
|
| esv3618680 | Chr.8:124089247 - 124098454 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map