Assay Details
Target Gene Details
Entrez Gene ID: | 89887 |
Gene Name: | zinc finger protein 628 |
Gene Aliases: |
ZEC, Zfp628 |
Location: |
Chr.19:55476332-55484487 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF628 | NM_033113.2 | 1 | 267 | NP_149104.3 |
| XM_005259371.3 | XP_005259428.1 | |||
| BC089449.1 | 1 | 267 | AAH89449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2552 | Chr.19:55467079 - 55498718 on Build GRCh38 | Insertion |
|
| nsv458833 | Chr.19:55466542 - 55521721 on Build GRCh38 | Loss |
|
| nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map