Assay Details
Target Gene Details
Entrez Gene ID: | 25953 |
Gene Name: | paroxysmal nonkinesigenic dyskinesia |
Gene Aliases: |
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2 |
Location: |
Chr.2:218270392-218346793 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNKD | NM_015488.4 | NP_056303.3 | ||
| XM_017003771.1 | XP_016859260.1 | |||
| AB033010.1 | BAA86498.1 | |||
| AK289867.1 | ||||
| BC036457.2 | AAH36457.1 |
Target Gene Details
Entrez Gene ID: | 64114 |
Gene Name: | transmembrane BAX inhibitor motif containing 1 |
Gene Aliases: |
LFG3, MST100, MSTP100, PP1201, RECS1 |
Location: |
Chr.2:218274192-218292577 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMBIM1 | NM_001321427.1 | NP_001308356.1 | ||
| NM_001321428.1 | NP_001308357.1 | |||
| NM_001321429.1 | NP_001308358.1 | |||
| NM_001321430.1 | NP_001308359.1 | |||
| NM_001321432.1 | NP_001308361.1 | |||
| NM_001321433.1 | NP_001308362.1 | |||
| NM_001321435.1 | NP_001308364.1 | |||
| NM_001321436.1 | NP_001308365.1 | |||
| NM_001321438.1 | NP_001308367.1 | |||
| NM_022152.5 | NP_071435.2 | |||
| NR_135643.1 | ||||
| XM_011511627.1 | XP_011509929.1 | |||
| AB257295.1 | ||||
| AF193045.1 | AAG22473.1 | |||
| AK027755.1 | BAB55346.1 | |||
| AK075465.1 | BAC11636.1 | |||
| AK090618.1 | ||||
| AK093911.1 | ||||
| AK130380.1 | ||||
| AK130713.1 | ||||
| AK298123.1 | ||||
| AK298500.1 | ||||
| AK300597.1 | ||||
| BC007980.2 | AAH07980.1 | |||
| BC013428.1 | AAH13428.1 | |||
| BC026348.1 | AAH26348.1 | |||
| BC026693.2 | AAH26693.1 | |||
| CF994144.1 |
More Information
Set Membership: |
|

Genomic Map