Assay Details
Target Gene Details
Entrez Gene ID: | 29843 |
Gene Name: | SUMO1/sentrin specific peptidase 1 |
Gene Aliases: |
SuPr-2 |
Location: |
Chr.12:48042893-48106308 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP1 | NM_001267594.1 | 18 | 2349 | NP_001254523.1 |
| NM_001267595.1 | 18 | 2604 | NP_001254524.1 | |
| NR_051991.1 | 19 | 2790 | ||
| NR_051992.1 | 19 | 2803 | ||
| XM_006719361.3 | 18 | 2484 | XP_006719424.1 | |
| XM_006719362.2 | 19 | 2256 | XP_006719425.1 | |
| XM_011538244.2 | 17 | 2485 | XP_011536546.1 | |
| XM_011538245.2 | 17 | 2436 | XP_011536547.1 | |
| XM_017019229.1 | 19 | 2622 | XP_016874718.1 | |
| XM_017019230.1 | 18 | 2538 | XP_016874719.1 | |
| XM_017019231.1 | 19 | 2522 | XP_016874720.1 | |
| XM_017019232.1 | 19 | 2249 | XP_016874721.1 | |
| XM_017019233.1 | 18 | 2532 | XP_016874722.1 | |
| XM_017019234.1 | 18 | 2442 | XP_016874723.1 | |
| XM_017019235.1 | 18 | 2175 | XP_016874724.1 | |
| XM_017019236.1 | 17 | 2410 | XP_016874725.1 | |
| XM_017019237.1 | 17 | 2310 | XP_016874726.1 | |
| XM_017019238.1 | 19 | 2560 | XP_016874727.1 | |
| XM_017019239.1 | 18 | 2600 | XP_016874728.1 | |
| XM_017019240.1 | 18 | 2521 | XP_016874729.1 | |
| AK292060.1 | 18 | 2120 | ||
| BC045639.1 | 18 | 2420 | AAH45639.2 | |
| BX537920.1 | 19 | 2803 | CAD97903.1 | |
| BX640784.1 | 19 | 2634 | CAE45874.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759898 | Chr.12:48035919 - 48431716 on Build GRCh38 | Gain+Loss |
|
| nsv832401 | Chr.12:47953670 - 48084216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map