Assay Details
Target Gene Details
Entrez Gene ID: | 56961 |
Gene Name: | Src homology 2 domain containing transforming protein D |
Gene Aliases: |
- |
Location: |
Chr.19:4278601-4290723 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHD | NM_020209.3 | NP_064594.3 | ||
| AK055673.1 | BAB70981.1 | |||
| AK056268.1 | ||||
| AL390078.1 | CAB98202.1 | |||
| BC007206.1 | AAH07206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643494 | Chr.19:4275790 - 4289414 on Build GRCh38 | Loss |
|
| nsv578442 | Chr.19:4236999 - 4312106 on Build GRCh38 | Loss |
|
| nsv833717 | Chr.19:4204757 - 4368519 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map