Assay Details
Target Gene Details
Entrez Gene ID: | 10590 |
Gene Name: | secretagogin, EF-hand calcium binding protein |
Gene Aliases: |
CALBL, DJ501N12.8, SECRET, SEGN, setagin |
Location: |
Chr.6:25652201-25701783 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCGN | NM_006998.3 | NP_008929.2 | ||
| XM_017010181.1 | XP_016865670.1 | |||
| AF420280.1 | AAL16052.1 | |||
| AK289477.1 | ||||
| BC000336.2 | AAH00336.1 | |||
| BC003036.1 | AAH03036.1 | |||
| BM511122.1 | ||||
| Y16752.1 | CAA76365.1 |
More Information
Set Membership: |
|

Genomic Map