Assay Details
Target Gene Details
Entrez Gene ID: | 6175 |
Gene Name: | ribosomal protein lateral stalk subunit P0 |
Gene Aliases: |
L10E, LP0, P0, PRLP0, RPP0 |
Location: |
Chr.12:120196700-120201211 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPLP0 | NM_001002.3 | NP_000993.1 | ||
| NM_053275.3 | NP_444505.1 | |||
| AK001313.1 | 1 | 244 | ||
| AK129754.1 | ||||
| AK129823.1 | ||||
| AK222571.1 | BAD96291.1 | |||
| AK291109.1 | ||||
| AK304674.1 | ||||
| BC003655.2 | AAH03655.1 | |||
| BC008092.1 | AAH08092.1 | |||
| BC015173.1 | AAH15173.1 | |||
| D28418.1 | BAA05784.1 | |||
| M17885.1 | AAA36470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976653 | Chr.12:120200045 - 120201198 on Build GRCh38 | Duplication |
|
| nsv832530 | Chr.12:120148265 - 120320372 on Build GRCh38 | Loss |
|
| nsv519657 | Chr.12:120095892 - 120238287 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map