Assay Details
Target Gene Details
Entrez Gene ID: | 90379 |
Gene Name: | DDB1 and CUL4 associated factor 15 |
Gene Aliases: |
C19orf72 |
Location: |
Chr.19:13952507-13961444 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF15 | NM_138353.2 | NP_612362.2 | ||
| AK093072.1 | ||||
| BC002926.2 | AAH02926.2 | |||
| BC013280.2 | AAH13280.2 | |||
| BC080575.1 | AAH80575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953979 | Chr.19:13795787 - 14006888 on Build GRCh38 | Deletion |
|
| nsv828451 | Chr.19:13937797 - 13965186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map