Assay Details
Target Gene Details
Entrez Gene ID: | 14 |
Gene Name: | angio associated migratory cell protein |
Gene Aliases: |
- |
Location: |
Chr.2:218264129-218270209 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAMP | NM_001087.4 | 11 | 1684 | NP_001078.2 |
| NM_001302545.1 | 11 | 1687 | NP_001289474.1 | |
| AB209790.1 | 8 | 2123 | BAD93027.1 | |
| AK131047.1 | 11 | 1621 | ||
| AK225078.1 | 11 | 1592 | ||
| BC008809.2 | 8 | 1167 | AAH08809.2 | |
| BC020244.1 | 11 | 1548 | AAH20244.1 | |
| BC039866.1 | 11 | 1612 | AAH39866.2 | |
| HY374777.1 | 1 | 146 | ||
| M95627.1 | 11 | 1615 | AAA68889.1 |
More Information
Set Membership: |
|

Genomic Map