Assay Details
Target Gene Details
Entrez Gene ID: | 8036 |
Gene Name: | SHOC2, leucine rich repeat scaffold protein |
Gene Aliases: |
SIAA0862, SOC2, SUR8 |
Location: |
Chr.10:110919370-111013667 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHOC2 | NM_001269039.2 | 2 | 342 | NP_001255968.1 |
| NM_001324336.1 | 2 | 299 | NP_001311265.1 | |
| NM_001324337.1 | 3 | 527 | NP_001311266.1 | |
| NM_007373.3 | 2 | 342 | NP_031399.2 | |
| NR_136749.1 | ||||
| XM_017016702.1 | 2 | 247 | XP_016872191.1 | |
| XM_017016703.1 | 2 | 6064 | XP_016872192.1 | |
| XM_017016704.1 | 2 | 246 | XP_016872193.1 | |
| AB020669.1 | 2 | 270 | ||
| AF054828.1 | 2 | 264 | AAC25698.1 | |
| AF068920.1 | AAC39856.1 | |||
| AK292833.1 | 3 | 468 | ||
| AK293271.1 | ||||
| BC050445.1 | 1 | 197 | AAH50445.1 | |
| BX649055.1 | 2 | 268 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035422 | Chr.10:110964401 - 111004836 on Build GRCh38 | Loss |
|
| nsv1035602 | Chr.10:110876295 - 111021655 on Build GRCh38 | Loss |
|
| esv3624545 | Chr.10:110948631 - 110987680 on Build GRCh38 | Loss |
|
| nsv831985 | Chr.10:110892331 - 111043345 on Build GRCh38 | Gain+Loss |
|
| esv2674974 | Chr.10:110948807 - 110987520 on Build GRCh38 | Deletion |
|
| dgv974n100 | Chr.10:110924064 - 111035401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map