Assay Details
Target Gene Details
Entrez Gene ID: | 27430 |
Gene Name: | methionine adenosyltransferase 2B |
Gene Aliases: |
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR |
Location: |
Chr.5:163503064-163519353 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAT2B | NM_013283.4 | 1 | 87 | NP_037415.1 |
| NM_182796.2 | NP_877725.1 | |||
| AB073390.1 | BAE45720.1 | |||
| AJ243721.1 | CAB56837.1 | |||
| AK096178.1 | ||||
| AK291919.1 | ||||
| AK312365.1 | ||||
| AL136664.1 | CAB66599.1 | |||
| AU279489.1 | ||||
| AW966728.1 | ||||
| AY358695.1 | AAQ89058.1 | |||
| BC005218.1 | AAH05218.1 | |||
| BC066645.1 | AAH66645.1 | |||
| BC093030.1 | AAH93030.1 | |||
| DA146198.1 | 1 | 87 | ||
| DQ395260.1 | ABD59011.1 | |||
| DQ413183.1 | ABD85290.1 | |||
| HY091756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017056 | Chr.5:163328870 - 163804011 on Build GRCh38 | Gain |
|
| nsv830538 | Chr.5:163377440 - 163541647 on Build GRCh38 | Gain+Loss |
|
| nsv1029437 | Chr.5:163104166 - 164109116 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map