Assay Details
Target Gene Details
| Entrez Gene ID: | 168400 | 
| Gene Name: | DEAD-box helicase 53 | 
| Gene Aliases: | CAGE, CT26 | 
| Location: | Chr.X:22999961-23002089 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DDX53 | NM_182699.3 | 1 | 1194 | NP_874358.2 | 
| AY039237.1 | 1 | 1199 | AAK72102.1 | |
| BC067878.1 | 1 | 1185 | AAH67878.1 | 
Target Gene Details
| Entrez Gene ID: | 100873065 | 
| Gene Name: | PTCHD1 antisense RNA (head to head) | 
| Gene Aliases: | DDX53-AS1, PTCHD1AS1, PTCHD1AS2 | 
| Location: | Chr.X:22193005-23293146 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PTCHD1-AS | NR_073010.2 | |||
| BX115199.1 | ||||
| DA355362.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3573648 | Chr.X:22953327 - 23080815 on Build GRCh38 | Loss |  PTCHD1-AS  DDX53 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top