Assay Details
Target Gene Details
Entrez Gene ID: | 1630 |
Gene Name: | DCC netrin 1 receptor |
Gene Aliases: |
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1 |
Location: |
Chr.18:52340172-53535903 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCC | NM_005215.3 | 1 | 398 | NP_005206.2 |
| XM_011525843.1 | 1 | 398 | XP_011524145.1 | |
| XM_017025568.1 | 1 | 398 | XP_016881057.1 | |
| XM_017025569.1 | 1 | 398 | XP_016881058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953545 | Chr.18:52339631 - 52343030 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map