Assay Details
Target Gene Details
Entrez Gene ID: | 83543 |
Gene Name: | allograft inflammatory factor 1 like |
Gene Aliases: |
C9orf58, IBA2 |
Location: |
Chr.9:131096476-131123152 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIF1L | NM_001185095.1 | NP_001172024.1 | ||
| NM_001185096.1 | NP_001172025.1 | |||
| NM_031426.3 | NP_113614.1 | |||
| NR_033701.1 | ||||
| AK022845.1 | BAB14269.1 | |||
| AK057789.1 | ||||
| AK091912.1 | BAC03770.1 | |||
| AK128526.1 | BAC87480.1 | |||
| AK314600.1 | ||||
| AL136566.1 | CAB66501.1 | |||
| AL833896.1 | CAD38752.1 | |||
| AY359067.1 | AAQ89426.1 | |||
| BC021253.2 | AAH21253.1 | |||
| BI766795.1 | ||||
| DA285481.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain |
|
| nsv831737 | Chr.9:130895103 - 131128557 on Build GRCh38 | Loss |
|
| nsv831738 | Chr.9:130976552 - 131153174 on Build GRCh38 | Loss |
|
| esv2759717 | Chr.9:130862179 - 131153174 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map