Assay Details
Target Gene Details
Entrez Gene ID: | 55818 |
Gene Name: | lysine demethylase 3A |
Gene Aliases: |
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA |
Location: |
Chr.2:86440647-86492716 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM3A | NM_001146688.1 | 10 | 1196 | NP_001140160.1 |
| NM_018433.5 | 10 | 1465 | NP_060903.2 | |
| XM_006712051.3 | 9 | 1168 | XP_006712114.1 | |
| XM_017004493.1 | 10 | 1484 | XP_016859982.1 | |
| XM_017004494.1 | 11 | 1307 | XP_016859983.1 | |
| AB018285.2 | 10 | 1228 | ||
| AK304276.1 | 8 | 982 | ||
| AL832150.1 | 10 | 1231 | CAH18459.3 | |
| BX640698.1 | 10 | 1266 | CAE45820.1 | |
| BX647271.1 | 10 | 1234 | ||
| BX647752.1 | 10 | 1294 | ||
| BX648841.1 | 10 | 1197 | ||
| BX649109.1 | 10 | 1235 | ||
| CR749581.1 | 10 | 1233 | CAH18373.1 |
More Information
Set Membership: |
|

Genomic Map