Assay Details
Target Gene Details
Entrez Gene ID: | 7326 |
Gene Name: | ubiquitin conjugating enzyme E2 G1 |
Gene Aliases: |
E217K, UBC7, UBE2G |
Location: |
Chr.17:4269217-4366674 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2G1 | NM_003342.4 | 6 | 3703 | NP_003333.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065237 | Chr.17:4040089 - 4278208 on Build GRCh38 | Gain |
|
| nsv1058176 | Chr.17:4189086 - 4502476 on Build GRCh38 | Gain |
|
| nsv1066651 | Chr.17:4269648 - 4303948 on Build GRCh38 | Gain |
|
| esv2762544 | Chr.17:4178680 - 4476552 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map