Assay Details
Target Gene Details
Entrez Gene ID: | 5981 |
Gene Name: | replication factor C subunit 1 |
Gene Aliases: |
A1, MHCBFB, PO-GA, RECC1, RFC, RFC140 |
Location: |
Chr.4:39287449-39366381 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFC1 | NM_001204747.1 | 17 | 2433 | NP_001191676.1 |
| NM_002913.4 | 17 | 2430 | NP_002904.3 | |
| XM_011513730.1 | 16 | 2355 | XP_011512032.1 | |
| XM_011513731.1 | 15 | 1923 | XP_011512033.1 | |
| XM_017008528.1 | 16 | 2352 | XP_016864017.1 | |
| AF040250.1 | 17 | 2335 | AAB99788.1 | |
| AK291612.1 | 17 | 2414 | ||
| BC035297.1 | 17 | 2384 | AAH35297.1 | |
| BC051751.1 | 17 | 2400 | AAH51751.1 | |
| BC051786.2 | 17 | 2414 | AAH51786.1 | |
| L23320.1 | 17 | 2499 | AAA16121.1 |
More Information
Set Membership: |
|

Genomic Map