Assay Details
Target Gene Details
Entrez Gene ID: | 1969 |
Gene Name: | EPH receptor A2 |
Gene Aliases: |
ARCC2, CTPA, CTPP1, CTRCT6, ECK |
Location: |
Chr.1:16124337-16156104 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA2 | XM_017000534.1 | XP_016856023.1 | ||
| XM_017000535.1 | XP_016856024.1 | |||
| XM_017000536.1 | XP_016856025.1 | |||
| AK296788.1 | ||||
| BC008655.1 | AAH08655.1 | |||
| BC037166.2 | AAH37166.1 | |||
| M59371.1 | AAA53375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833869 | Chr.1:16110258 - 16278634 on Build GRCh38 | Loss |
|
| nsv524045 | Chr.1:16005414 - 16226277 on Build GRCh38 | Loss |
|
| nsv508936 | Chr.1:15915852 - 16181025 on Build GRCh38 | Insertion |
|
| nsv833758 | Chr.1:15969028 - 16140519 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map