Assay Details
Target Gene Details
Entrez Gene ID: | 5330 |
Gene Name: | phospholipase C beta 2 |
Gene Aliases: |
PLC-beta-2 |
Location: |
Chr.15:40285496-40307973 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCB2 | NM_001284297.1 | NP_001271226.1 | ||
| NM_001284298.1 | NP_001271227.1 | |||
| NM_004573.2 | NP_004564.2 | |||
| XM_011521674.2 | XP_011519976.1 | |||
| XM_017022314.1 | XP_016877803.1 | |||
| XM_017022315.1 | XP_016877804.1 | |||
| XM_017022316.1 | XP_016877805.1 | |||
| XM_017022317.1 | XP_016877806.1 | |||
| XM_017022318.1 | XP_016877807.1 | |||
| XM_017022319.1 | XP_016877808.1 | |||
| AB209583.1 | BAD92820.1 | |||
| AK130831.1 | BAC85441.1 | |||
| AK291657.1 | ||||
| BC136467.1 | ||||
| M95678.1 | AAA36453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471240 | Chr.15:40235954 - 40303426 on Build GRCh38 | Loss |
|
| nsv510660 | Chr.15:40260705 - 40429684 on Build GRCh38 | Deletion |
|
| nsv569227 | Chr.15:40273504 - 40314283 on Build GRCh38 | Loss |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
| nsv517541 | Chr.15:40265067 - 40303426 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map