Assay Details
Target Gene Details
Entrez Gene ID: | 9950 |
Gene Name: | golgin A5 |
Gene Aliases: |
GOLIM5, RFG5, ret-II |
Location: |
Chr.14:92794231-92839963 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA5 | NM_005113.3 | 6 | 1447 | NP_005104.3 |
| XM_011537420.2 | 6 | 1447 | XP_011535722.1 | |
| AF085199.1 | 6 | 1322 | AAD09753.1 | |
| AJ132949.1 | 5 | 1191 | CAB36967.1 | |
| AK293077.1 | 6 | 1350 | ||
| AK303789.1 | 7 | 1080 | ||
| AY644768.1 | 5 | 1227 | AAV85456.1 | |
| BC023021.1 | 6 | 1355 | AAH23021.1 |
More Information
Set Membership: |
|

Genomic Map