Assay Details
Target Gene Details
Entrez Gene ID: | 9125 |
Gene Name: | CCR4-NOT transcription complex subunit 9 |
Gene Aliases: |
CAF40, CT129, RCD-1, RCD1, RQCD1 |
Location: |
Chr.2:218568579-218596435 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT9 | NM_001271634.1 | 7 | 1053 | NP_001258563.1 |
| NM_001271635.1 | 6 | 957 | NP_001258564.1 | |
| NM_005444.2 | 6 | 957 | NP_005435.1 | |
| NR_073390.1 | 5 | 847 | ||
| XM_011512138.2 | 7 | 1076 | XP_011510440.1 | |
| XM_017005248.1 | 6 | 1546 | XP_016860737.1 | |
| XM_017005249.1 | 7 | 1078 | XP_016860738.1 | |
| AB500892.1 | 6 | 829 | ||
| AK293281.1 | 7 | 1053 | ||
| AK296037.1 | 2 | 110 | ||
| AK316567.1 | 6 | 663 | ||
| BC007102.1 | 6 | 664 | ||
| BC137455.1 | 6 | 679 | ||
| BC137456.1 | 6 | 679 |
More Information
Set Membership: |
|

Genomic Map