Assay Details
Target Gene Details
Entrez Gene ID: | 10518 |
Gene Name: | calcium and integrin binding family member 2 |
Gene Aliases: |
DFNB48, KIP2, USH1J |
Location: |
Chr.15:78104606-78131976 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB2 | NM_001271888.1 | 5 | 1481 | NP_001258817.1 |
| NM_001271889.1 | 4 | 1369 | NP_001258818.1 | |
| NM_001301224.1 | 5 | 1531 | NP_001288153.1 | |
| NM_006383.3 | 6 | 1516 | NP_006374.1 | |
| NR_125435.1 | 5 | 1404 | ||
| XM_006720374.2 | XP_006720437.1 | |||
| AA731290.1 | 1 | 109 | ||
| AB012955.1 | BAA33584.1 | |||
| BC033108.1 | ||||
| BC047381.1 | AAH47381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv274890 | Chr.15:78104105 - 78109916 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map