Assay Details
Target Gene Details
Entrez Gene ID: | 4920 |
Gene Name: | receptor tyrosine kinase like orphan receptor 2 |
Gene Aliases: |
BDB, BDB1, NTRKR2 |
Location: |
Chr.9:91722596-91950206 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ROR2 | NM_001318204.1 | 8 | 1452 | NP_001305133.1 |
| NM_004560.3 | 8 | 1486 | NP_004551.2 | |
| XM_005252008.4 | 8 | 1451 | XP_005252065.1 | |
| XM_005252009.3 | 2 | 192 | XP_005252066.1 | |
| XM_006717121.3 | 8 | 2108 | XP_006717184.1 | |
| XM_017014762.1 | 8 | 3183 | XP_016870251.1 | |
| XM_017014763.1 | 8 | 1362 | XP_016870252.1 | |
| AB209154.1 | 10 | 1755 | BAD92391.1 | |
| BC130522.1 | 8 | 1374 | ||
| BI089313.1 | ||||
| M97639.1 | 8 | 1486 | AAA60276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045332 | Chr.9:91590379 - 91893734 on Build GRCh38 | Loss |
|
| esv3576683 | Chr.9:91651711 - 91758339 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map